missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Thioredoxin-2 Protein

A DNA sequence encoding the Escherichia coli (strain K12) Thioredoxin-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7012-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16057030

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_417077.1
Thioredoxin-2 Protein
200 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
947062
Greater than 90% as determined by SDS-PAGE.
Research Use Only
trxC
Recombinant Protein
E. coli
MNTVCTHCQA INRIPDDRIE DAAKCGRCGH DLFDGEVINA TGETLDKLLK DDLPVVIDFW APWCGPCRNF APIFEDVAQE RSGKVRFVKV NTEAERELSS RFGIRSIPTI MIFKNGQVVD MLNGAVPKAP FDSWLNESL
Videos
SDS
Documents