missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Thioredoxin-2 Protein
A DNA sequence encoding the Escherichia coli (strain K12) Thioredoxin-2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7012-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_417077.1 | |
Thioredoxin-2 Protein | |
Research Use Only | |
trxC | |
Recombinant Protein | |
E. coli | |
MNTVCTHCQA INRIPDDRIE DAAKCGRCGH DLFDGEVINA TGETLDKLLK DDLPVVIDFW APWCGPCRNF APIFEDVAQE RSGKVRFVKV NTEAERELSS RFGIRSIPTI MIFKNGQVVD MLNGAVPKAP FDSWLNESL | |
Greater than 90% as determined by SDS-PAGE. |
947062 | |
1 mg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |