missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Thiosulfate sulfurtransferase GlpE Protein
A DNA sequence encoding the Escherichia coli (strain ATCC 8739 / DSM 1576 / Crooks) Thiosulfate sulfurtransferase GlpE, was expressed in the hosts and tags indicated.
3490.00 SEK - 32610.00 SEK
Specifications
Accession Number | WP_000371929.1 |
---|---|
Name | Thiosulfate sulfurtransferase GlpE Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | glpE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16049060
|
enQuireBio™
QP7505-YE-100UG |
100 μg |
12410.00 SEK
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16089060
|
enQuireBio™
QP7505-YE-500UG |
500 μg |
29995.00 SEK
500µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16059060
|
enQuireBio™
QP7505-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079060
|
enQuireBio™
QP7505-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099060
|
enQuireBio™
QP7505-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069060
|
enQuireBio™
QP7505-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
WP_000371929.1 | |
Research Use Only | |
glpE | |
Recombinant Protein | |
Yeast | |
MDQFECINVA DAHQKLQEKE AVLVDIRDPQ SFAMGHAVQA FHLTNDTLGA FMRDNDFDTP VMVMCYHGNS SKGAAQYLLQ QGYDVVYSID GGFEVWQRQF PAEVAYGA | |
Greater than 90% as determined by SDS-PAGE. |
Thiosulfate sulfurtransferase GlpE Protein | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |