missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Trigger factor Protein
A DNA sequence encoding the Escherichia coli O127:H6 (strain E2348/69 / EPEC) Trigger factor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7443-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
WP_001198386.1 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |
Trigger factor Protein | |
Research Use Only | |
tig | |
Recombinant Protein | |
Yeast | |
MQVSVETTQG LGRRVTITIA ADSIETAVKS ELVNVAKKVR IDGFRKGKVP MNIVAQRYGA SVRQDVLGDL MSRNFIDAII KEKINPAGAP TYVPGEYKLG EDFTYSVEFE VYPEVELQGL EAIEVEKPIV EVTDADVDGM LDTLRKQQAT WKEKDGAVEA EDRVTIDFTG SVDGEEFEGG KASDFVLAMG QGRMIPGFED GIKGHKAGEE FTIDVTFPEE YHAENLKGKA AKFAINLKKV EERELPELTA EFIKRFGVED GSVEGLRAEV RKNMERELKS AIRNRVKSQA IEGLVKANDI DVPAALIDSE IDVLRRQAAQ RFGGNEKQAL ELPRELFEEQ AKRRVVVGLL LGEVIRTNEL KADEERVKGL IEEMASAYED PKEVIEFYSK NKELMDNMRN VALEEQAVEA VLAKAKVTEK ETTFNELMNQ QA | |
Greater than 90% as determined by SDS-PAGE. |