Learn More
enQuireBio™ Recombinant Epstein-Barr Envelope glycoprotein 1
A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Envelope glycoprotein H, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7411-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
YP_401700.1 | |
Envelope glycoprotein 1 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Epstein-Barr | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3783742 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
gH | |
Recombinant Protein | |
E. coli | |
SLSEVKLHLD IEGHASHYTI PWTELMAKVP GLSPEALWRE ANVTEDLASM LNRYKLIYKT SGTLGIALAE PVDIPAVSEG SMQVDASKVH PGVISGLNSP ACMLSAPLEK QLFYYIGTML PNTRPHSYVF YQLRCHLSYV ALSINGDKFQ YTGAMTSKFL MGTYKRVTEK GDEHVLSLVF GKTKDLPDLR GPFSYPSLTS |