missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-2 gp-36 397aa Protein
A cDNA sequence encoding the HIV-2 gp-36 397aa was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12270-500ug
Additional Details : Weight : 0.01000kg
Specifications
HIV-2 gp-36 397aa Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
EQTMVQDDPSTCRGEFLYCNMTWFLNWIENKTHRNYAPCHIKQIINTWHKVGRNVYLPPREGELSCNSTVTSIIANIDWQNNNQTNITFSAEVAELYRLELGDYKLVEITPIGFAPTKEKRYSSAHGRHTRGVFVLGFLGFLATAGSAMGAASLTVSAQSRTLLAGIVQQQQQLLDVVKRQQELLRLTVWGTKNLQARVTAIEKYLQDQARLNSWGCAFRQVCHTTVPWVNDSLAPDWDNMTWQEWEKQVRYLEANISKSLEQAQIQQEKNMYELQKLNSWDIFGNWFDLTSWVKYIQYGVLIIVAVIALRIVIYVVQMLSRLRKGYRPVFSSPPGYIQQIHIHKDRGQPANEETEEDGGSNGGDRYWPWPIAYIHFLIRQLIRLLTRLYSICSQAC | |
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE. |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
0.01M Na2CO3, 0.01M Na3EDTA, 0.014 Mb-mercaptoethanol, 0.05% Tween-20. |