Learn More
enQuireBio™ Recombinant HIV-2 gp-36 397aa Protein
A cDNA sequence encoding the HIV-2 gp-36 397aa was constructed and used to recombinantly synthesize the protein.
Brand:  enQuireBio™ QP12270-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15999888
  • 20560.00 SEK / 1mg
Estimated Shipment: 28-06-2024
to see stock.
Specifications
HIV-2 gp-36 397aa Protein
Research Use Only
Recombinant Protein
E. coli
EQTMVQDDPSTCRGEFLYCNMTWFLNWIENKTHRNYAPCHIKQIINTWHKVGRNVYLPPREGELSCNSTVTSIIANIDWQNNNQTNITFSAEVAELYRLELGDYKLVEITPIGFAPTKEKRYSSAHGRHTRGVFVLGFLGFLATAGSAMGAASLTVSAQSRTLLAGIVQQQQQLLDVVKRQQELLRLTVWGTKNLQARVTAIEKYLQDQARLNSWGCAFRQVCHTTVPWVNDSLAPDWDNMTWQEWEKQVRYLEANISKSLEQAQIQQEKNMYELQKLNSWDIFGNWFDLTSWVKYIQYGVLIIVAVIALRIVIYVVQMLSRLRKGYRPVFSSPPGYIQQIHIHKDRGQPANEETEEDGGSNGGDRYWPWPIAYIHFLIRQLIRLLTRLYSICSQAC
Greater than 95.0% as determined by HPLC analysis and SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
Untagged
0.01M Na2CO3, 0.01M Na3EDTA, 0.014 Mb-mercaptoethanol, 0.05% Tween-20.