missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV-2 gp36 Protein
A cDNA sequence encoding the HIV-2 gp36 was constructed and used to recombinantly synthesize the protein.
2405.00 SEK - 10265.00 SEK
Specifications
Name | HIV-2 gp36 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HIV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959888
|
enQuireBio™
QP12269-HIS-100UG |
100 μg |
2405.00 SEK
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979888
|
enQuireBio™
QP12269-HIS-500UG |
500 μg |
6835.00 SEK
500µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15969888
|
enQuireBio™
QP12269-HIS-1MG |
1 mg |
10265.00 SEK
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HIV-2 gp36 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
His | |
Greater than 95.0% as determined by HPLC. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
(1 mg/1ml) in H2O. |