missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Horse Latherin Protein

A DNA sequence encoding the Equus caballus (Horse) Latherin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9489-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16058421

  • 20629.00 SEK / 500µg


Estimated Shipment
03-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_001075328.2
Latherin Protein
500 μg
Not determined.
Full Length of Mature Protein
Horse
His
Tris-based buffer, 50% glycerol
100033921
Greater than 86.7% as determined by SDS-PAGE.
Research Use Only
LATH
Recombinant Protein
Yeast
QQIPPEVSSQ ITDALTQGLL DGNFLSLLNA INLEGLLNTI LDQVTGLLNI LVGPLLGPSD AEIKLQDTRL LQLSLEFSPD SKGIDIWIPL ELSVYLKLLI LEPLTLYVRT DIRVQLRLES DEDGKYRLAF GHCSLLPRAI ELQSGNPLSL PVNAVLGTIE NALGNFITED LGAGLCPTLN SLVSNLDLQL VNNLINLILD RANVDLSV
Videos
SDS
Documents