missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HCV Core 169aa Protein
A cDNA sequence encoding the HCV Core 169aa was constructed and used to recombinantly synthesize the protein.
2690.00 SEK - 20560.00 SEK
Specifications
Name | HSV HCV Core 169aa Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | HSV |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959498
|
enQuireBio™
QP12141-100UG |
100 μg |
2690.00 SEK
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15979498
|
enQuireBio™
QP12141-500UG |
500 μg |
10265.00 SEK
500µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15969498
|
enQuireBio™
QP12141-1MG |
1 mg |
20560.00 SEK
1mg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HSV HCV Core 169aa Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
His | |
Greater than 95.0% as determined by SDS-PAGE. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Sterile filtered solution containing Phosphate saline buffer, 50mM arginine and 1M urea. |