missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HSV HSV-1 gD Protein
A cDNA sequence encoding the HSV-1 gD was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12339-500ug
Additional Details : Weight : 0.01000kg
Specifications
HSV HSV-1 gD Protein | |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
GST |
HSV-1 gD protein is >95% pure as determined by 10% PAGE (coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl pH 8.0, 1mM EDTA, and 50% glycerol. |