missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 14-3-3 tau / 14-3-3 theta / YWHAQ Protein
A DNA sequence encoding the Homo sapiens (Human) 14-3-3 tau / 14-3-3 theta / YWHAQ, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP873-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_006817.1 | |
14-3-3 tau / 14-3-3 theta / YWHAQ Protein | |
Research Use Only | |
YWHAQ | |
Recombinant Protein | |
E. coli | |
MEKTELIQKA KLAEQAERYD DMATCMKAVT EQGAELSNEE RNLLSVAYKN VVGGRRSAWR VISSIEQKTD TSDKKLQLIK DYREKVESEL RSICTTVLEL LDKYLIANAT NPESKVFYLK MKGDYFRYLA EVACGDDRKQ TIDNSQGAYQ EAFDISKKEM QPTHPIRLGL ALNFSVFYYE ILNNPELACT LAKTAFDEAI AELDTLNEDS YKDSTLIMQL LRDNLTLWTS DSAGEECDAA EGAEN | |
Greater than 80% as determined by SDS-PAGE. |
10971 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |