missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 28S ribosomal protein S16, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 28S ribosomal protein S16, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8290-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_057149.1 | |
28S ribosomal protein S16, mitochondrial Protein | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
51021 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
MRPS16 | |
Recombinant Protein | |
E. coli | |
VAAHNKCPRD GRFVEQLGSY DPLPNSHGEK LVALNLDRIR HWIGCGAHLS KPMEKLLGLA GFFPLHPMMI TNAERLRRKR AREVLLASQK TDAEATDTEA TET |