missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 3-hydroxyacyl-CoA dehydrogenase type-2 Protein
A DNA sequence encoding the Homo sapiens (Human) 3-hydroxyacyl-CoA dehydrogenase type-2, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001032900.1 |
---|---|
Gene ID (Entrez) | 3028 |
Name | 3-hydroxyacyl-CoA dehydrogenase type-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16062301
|
enQuireBio™
QP8037-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16082301
|
enQuireBio™
QP8037-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16072301
|
enQuireBio™
QP8037-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16092301
|
enQuireBio™
QP8037-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16002311
|
enQuireBio™
QP8037-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012311
|
enQuireBio™
QP8037-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001032900.1 | |
3-hydroxyacyl-CoA dehydrogenase type-2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3028 | |
Research Use Only | |
HSD17B10 | |
Recombinant Protein | |
E. coli | |
AAACRSVKGL VAVITGGASG LGLATAERLV GQGASAVLLD LPNSGGEAQA KKLGNNCVFA PADVTSEKDV QTALALAKGK FGRVDVAVNC AGIAVASKTY NLKKGQTHTL EDFQRVLDVN LMGTFNVIRL VAGEMGQNEP DQGGQRGVII NTASVAAFEG QVGQAAYSAS KGGIVGMTLP IARDLAPIGI RVMTIAPGLF GTPLLTSLPE KVCNFLASQV PFPSRLGDPA EYAHLVQAII ENPFLNGEVI RLDGAIRMQP | |
Greater than 90% as determined by SDS-PAGE. |