Läs mer
enQuireBio™ Recombinant Human 39S ribosomal protein L12, mitochondrial Protein
Produktkod. 16032900
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Förpackningsstorlek:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16032900 50 μg 50µg
16092890 10 μg 10µg
16082890 100 μg 100µg
16012900 200 μg 200µg
16022900 500 μg 500µg
16002900 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Produktkod. 16032900 Leverantör enQuireBio™ Leverantörsnummer QP6380ec50ug
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L12, mitochondrial, was expressed in the hosts and tags indicated.
Specifikationer
Tillträdesnummer NP_002940.2
Gen-ID (Entrez) 6182
Namn 39S ribosomal protein L12, mitochondrial Protein
Kvantitet 50 μg
Regulatorisk status Research Use Only
Endotoxinkoncentration Not determined.
Gensymbol MRPL12
Protein Längd Full Length
Produkttyp Recombinant Protein
Korsreaktivitet Human
Art E. coli
Protein Tag GST
Sekvens EALAGAPLDN APKEYPPKIQ QLVQDIASLT LLEISDLNEL LKKTLKIQDV GLVPMGGVMS GAVPAAAAQE AVEEDIPIAK ERTHFTVRLT EAKPVDKVKL IKEIKNYIQG INLVQAKKLV ESLPQEIKAN VAKAEAEKIK AALEAVGGTV VLE
Buffert Tris-based buffer, 50% glycerol
Renhet eller kvalitetsklass Greater than 80% as determined by SDS-PAGE.