missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 39S ribosomal protein L19, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L19, mitochondrial, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_055578.2 |
---|---|
Gene ID (Entrez) | 9801 |
Name | 39S ribosomal protein L19, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16042900
|
enQuireBio™
QP6381-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16062900
|
enQuireBio™
QP6381-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16052900
|
enQuireBio™
QP6381-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072900
|
enQuireBio™
QP6381-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16082900
|
enQuireBio™
QP6381-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16092900
|
enQuireBio™
QP6381-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_055578.2 | |
39S ribosomal protein L19, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
9801 | |
Research Use Only | |
MRPL19 | |
Recombinant Protein | |
E. coli | |
MAACIAAGHW AAMGLGRSFQ AARTLLPPPA SIACRVHAGP VRQQSTGPSE PGAFQPPPKP VIVDKHRPVE PERRFLSPEF IPRRGRTDPL KFQIERKDML ERRKVLHIPE FYVGSILRVT TADPYASGKI SQFLGICIQR SGRGLGATFI LRNVIEGQGV EICFELYNPR VQEIQVVKLE KRLDDSLLYL RDALPEYSTF DVNMKPVVQE PNQKVPVNEL KVKMKPKPWS KRWERPNFNI KGIRFDLCLT EQQMKEAQKW NQPWLEFDMM REYDTSKIEA AIWKEIEASK RS | |
Greater than 90% as determined by SDS-PAGE. |