missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 39S ribosomal protein L54, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L54, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6383-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_758455.1 | |
39S ribosomal protein L54, mitochondrial Protein | |
Research Use Only | |
MRPL54 | |
Recombinant Protein | |
E. coli | |
GWGAWELLNP ATSGRLLARD YAKKPVMKGA KSGKGAVTSE ALKDPDVCTD PVQLTTYAMG VNIYKEGQDV PLKPDAEYPE WLFEMNLGPP KTLEELDPES REYWRRLRKQ NIWRHNRLSK NKRL | |
Greater than 90% as determined by SDS-PAGE. |
116541 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |