missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 39S ribosomal protein L55, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 39S ribosomal protein L55, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6384-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001308213.1 | |
39S ribosomal protein L55, mitochondrial Protein | |
Research Use Only | |
MRPL55 | |
Recombinant Protein | |
E. coli | |
DSSRASLTRV HRQAYARLYP VLLVKQDGST IHIRYREPRR MLAMPIDLDT LSPEERRARL RKREAQLQSR KEYEQELSDD LHVERYRQFW TRTKK | |
Greater than 90% as determined by SDS-PAGE. |
128308 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |