missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 40S ribosomal protein S18 Protein
A DNA sequence encoding the Homo sapiens (Human) 40S ribosomal protein S18, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_072045.1 |
---|---|
Gene ID (Entrez) | 6222 |
Name | 40S ribosomal protein S18 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16064311
|
enQuireBio™
QP8380-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16084311
|
enQuireBio™
QP8380-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16074311
|
enQuireBio™
QP8380-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094311
|
enQuireBio™
QP8380-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004321
|
enQuireBio™
QP8380-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014321
|
enQuireBio™
QP8380-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spezifikation
NP_072045.1 | |
40S ribosomal protein S18 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6222 | |
Research Use Only | |
RPS18 | |
Recombinant Protein | |
E. coli | |
SLVIPEKFQH ILRVLNTNID GRRKIAFAIT AIKGVGRRYA HVVLRKADID LTKRAGELTE DEVERVITIM QNPRQYKIPD WFLNRQKDVK DGKYSQVLAN GLDNKLREDL ERLKKIRAHR GLRHFWGLRV RGQHTKTTGR RGRTVGVSKK K | |
Greater than 90% as determined by SDS-PAGE. |