missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S acidic ribosomal protein P0 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S acidic ribosomal protein P0, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6623-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000993.1 | |
60S acidic ribosomal protein P0 Protein | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6175 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
RPLP0 | |
Recombinant Protein | |
E. coli | |
MPREDRATWK SNYFLKIIQL LDDYPKCFIV GADNVGSKQM QQIRMSLRGK AVVLMGKNTM MRKAIRGHLE NNPALEKLLP HIRGNVGFVF TKEDLTEIRD MLLANKVPAA ARAGAIAPCE VTVPAQNTGL GPEKTSFFQA LGITTKISRG TIEILSDVQL IKTGDKVGAS EATLLNMLNI SPFSFGLVIQ QVFDNGSIYN PEVLDITEET LHSRFLEGVR NVASVCLQIG YPTVASVPHS IINGYKRVLA LSVETDYTFP LAEKVKAFLA DPSAFVAAAP VAAATTAAPA AAAAPAKVEA KEESEESDED MGFGLFD |