missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S acidic ribosomal protein P2 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S acidic ribosomal protein P2, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_000995.1 |
---|---|
Gene ID (Entrez) | 6181 |
Name | 60S acidic ribosomal protein P2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044421
|
enQuireBio™
QP8404-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064421
|
enQuireBio™
QP8404-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054421
|
enQuireBio™
QP8404-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074421
|
enQuireBio™
QP8404-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084421
|
enQuireBio™
QP8404-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094421
|
enQuireBio™
QP8404-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000995.1 | |
60S acidic ribosomal protein P2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6181 | |
Research Use Only | |
RPLP2 | |
Recombinant Protein | |
E. coli | |
MRYVASYLLA ALGGNSSPSA KDIKKILDSV GIEADDDRLN KVISELNGKN IEDVIAQGIG KLASVPAGGA VAVSAAPGSA APAAGSAPAA AEEKKDEKKE ESEESDDDMG FGL | |
Greater than 90% as determined by SDS-PAGE. |