missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L18 Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L18, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_000970.1 |
---|---|
Gene ID (Entrez) | 6141 |
Name | 60S ribosomal protein L18 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044361
|
enQuireBio™
QP8388-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064361
|
enQuireBio™
QP8388-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054361
|
enQuireBio™
QP8388-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074361
|
enQuireBio™
QP8388-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084361
|
enQuireBio™
QP8388-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094361
|
enQuireBio™
QP8388-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000970.1 | |
60S ribosomal protein L18 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6141 | |
Research Use Only | |
RPL18 | |
Recombinant Protein | |
E. coli | |
GVDIRHNKDR KVRRKEPKSQ DIYLRLLVKL YRFLARRTNS TFNQVVLKRL FMSRTNRPPL SLSRMIRKMK LPGRENKTAV VVGTITDDVR VQEVPKLKVC ALRVTSRARS RILRAGGKIL TFDQLALDSP KGCGTVLLSG PRKGREVYRH FGKAPGTPHS HTKPYVRSKG RKFERARGRR ASRGYK | |
Greater than 90% as determined by SDS-PAGE. |