missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 60S ribosomal protein L36a-like Protein
A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L36a-like, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8385-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000992.1 | |
60S ribosomal protein L36a-like Protein | |
Research Use Only | |
RPL36AL | |
Recombinant Protein | |
E. coli | |
MVNVPKTRRT FCKKCGKHQP HKVTQYKKGK DSLYAQGRRR YDRKQSGYGG QTKPIFRKKA KTTKKIVLRL ECVEPNCRSK RMLAIKRCKH FELGGDKKRK GQVIQF | |
Greater than 90% as determined by SDS-PAGE. |
6166 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |