missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 60S ribosomal protein L9 Protein

A DNA sequence encoding the Homo sapiens (Human) 60S ribosomal protein L9, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6622-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064610

  • 6002.00 SEK / 100µg


Estimated Shipment
08-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_000652.2
60S ribosomal protein L9 Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
6133
Greater than 80% as determined by SDS-PAGE.
Research Use Only
RPL9
Recombinant Protein
E. coli
MKTILSNQTV DIPENVDITL KGRTVIVKGP RGTLRRDFNH INVELSLLGK KKKRLRVDKW WGNRKELATV RTICSHVQNM IKGVTLGFRY KMRSVYAHFP INVVIQENGS LVEIRNFLGE KYIRRVRMRP GVACSVSQAQ KDELILEGND IELVSNSAAL IQQATTVKNK DIRKFLDGIY VSEKGTVQQA DE
Videos
SDS
Documents