missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AADAT Protein
A DNA sequence encoding the Homo sapiens (Human) AADAT, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7920-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001273611.1 | |
AADAT Protein | |
Research Use Only | |
AADAT | |
Recombinant Protein | |
E. coli | |
PKSMISLAGG LPNPNMFPFK TAVITVENGK TIQFGEEMMK RALQYSPSAG IPELLSWLKQ LQIKLHNPPT IHYPPSQGQM DLCVTSGSQQ GLCKVFEMII NPGDNVLLDE PAYSGTLQSL HPLGCNIINV ASDESGIVPD SLRDILSRWK PEDAKNPQKN TPKFLYTVPN GNNPTGNSLT SERKKEIYEL ARKYDFLIIE DDPYYFLQFN KFRVPTFLSM DVDGRVIRAD SFSKIISSGL RIGFLTGPKP LIERVILHIQ VSTLHPSTFN QLMISQLLHE WGEEGFMAHV DRVIDFYSNQ KDAILAAADK WLTGLAEWHV PAAGMFLWIK VKGINDVKEL IEEKAVKMGV LMLPGNAFYV DSSAPSPYLR ASFSSASPEQ MDVAFQVLAQ LIKESL | |
Greater than 90% as determined by SDS-PAGE. |
51166 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |