missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Acid ceramidase Protein
A DNA sequence encoding the Homo sapiens (Human) Acid ceramidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7569-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001120977.1 | |
Acid ceramidase Protein | |
50 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
427 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
ASAH1 | |
Recombinant Protein | |
E. coli | |
QHAPPWTEDC RKSTYPPSGP TYRGAVPWYT INLDLPPYKR WHELMLDKAP VLKVIVNSLK NMINTFVPSG KIMQVVDEKL PGLLGNFPGP FEEEMKGIAA VTDIPLGEII SFNIFYELFT ICTSIVAEDK KGHLIHGRNM DFGVFLGWNI NNDTWVITEQ LKPLTVNLDF QRNNKTVFKA SSFAGYVGML TGFKPGLFSL TLNERFSING GYLGILEWIL GKKDVMWIGF LTRTVLENST SYEEAKNLLT KTKILAPAYF ILGGNQSGEG CVITRDRKES LDVYELDAKQ GRWYVVQTNY DRWKHPFFLD DRRTPAKMCL NRTSQENISF ETMYDVLSTK PVLNKLTVYT TLIDVTKGQF ETYLRDCPDP CIGW |