missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Acid ceramidase Protein

A DNA sequence encoding the Homo sapiens (Human) Acid ceramidase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7569-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16039460

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001120977.1
Acid ceramidase Protein
Research Use Only
ASAH1
Recombinant Protein
E. coli
QHAPPWTEDC RKSTYPPSGP TYRGAVPWYT INLDLPPYKR WHELMLDKAP VLKVIVNSLK NMINTFVPSG KIMQVVDEKL PGLLGNFPGP FEEEMKGIAA VTDIPLGEII SFNIFYELFT ICTSIVAEDK KGHLIHGRNM DFGVFLGWNI NNDTWVITEQ LKPLTVNLDF QRNNKTVFKA SSFAGYVGML TGFKPGLFSL TLNERFSING GYLGILEWIL GKKDVMWIGF LTRTVLENST SYEEAKNLLT KTKILAPAYF ILGGNQSGEG CVITRDRKES LDVYELDAKQ GRWYVVQTNY DRWKHPFFLD DRRTPAKMCL NRTSQENISF ETMYDVLSTK PVLNKLTVYT TLIDVTKGQF ETYLRDCPDP CIGW
Greater than 90% as determined by SDS-PAGE.
427
200 μg
Not determined.
Full Length of Mature Protein
Human
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers