missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ACP5 Protein

A cDNA sequence encoding the ACP5 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10502-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934068

  • 20560.00 SEK / 100µg
Estimated Shipment: 10-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

54
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is >10,000 units/mg, and is defined as the amount of enzyme that hydrolyze 1.0 nmoles of p-nitrophenyl phosphate (pNPP) per minute at pH 5.0 at 37C.
Human
Untagged
ACP5 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
ACP5 Protein
Research Use Only
ACP5
Recombinant Protein
Baculovirus
ATPALRFVAV GDWGGVPNAP FHTAREMANA KEIARTVQIL GADFILSLGD NFYFTGVQDI NDKRFQETFE DVFSDRSLRK VPWYVLAGNH DHLGNVSAQI AYSKISKRWN FPSPFYRLHF KIPQTNVSVA IFMLDTVTLC GNSDDFLSQQ PERPRDVKLA RTQLSWLKKQ LAAAREDYVL VAGHYPVWSI AEHGPTHCLV KQLRPLLATY GVTAYLCGHD HNLQYLQDEN GVGYVLSGAG NFMDPSKRHQ RKVPNGYLRF HYGTEDSLGG FAYVEISSKE MTVTYIEASG KSLFKTRLPR RARP
Greater than 90.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers