Läs mer
R&D Systems™ Recombinant Human Active ITK (aa 352-620) Protein
:
10ug, Carrier Free
Beskrivning
The Recombinant Human Active ITK (aa 352-620) Protein is derived from Sf 9 (baculovirus). The Recombinant Human Active ITK (aa 352-620) Protein has been validated for the following applications: Bioactivity.
Specifikationer
Specifikationer
| För användning med (applikation) | Bioactivity |
| Formulering | Supplied in 50mM Tris-HCl (pH 7.5), 150mM NaCl, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, and 25% glycerol. |
| Molekylvikt (g/mol) | Observed MW: 53kDa |
| Källa | Spodoptera frugiperda,Sf 9 (baculovirus)-derived human ITK protein aa 352-620 Accession &Num; NM_005546 |
| Förvaringskrav | This product is stable at ≤ -70° C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature. Avoid repeated freeze-thaw cycles. |
| Konjugera | Unconjugated |
| Renhet eller kvalitetsklass | >70%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5μg per lane |