missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACVR2B Protein
A DNA sequence encoding the Homo sapiens (Human) ACVR2B, was expressed in the hosts and tags indicated.
2005.00 SEK - 31280.00 SEK
Specifications
Accession Number | NP_001097.2 |
---|---|
Gene ID (Entrez) | 93 |
Name | ACVR2B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008691
|
enQuireBio™
QP9768-YE-100UG |
100 μg |
7265.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028691
|
enQuireBio™
QP9768-YE-1MG |
1 mg |
31280.00 SEK
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018691
|
enQuireBio™
QP9768-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038691
|
enQuireBio™
QP9768-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048691
|
enQuireBio™
QP9768-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058691
|
enQuireBio™
QP9768-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001097.2 | |
ACVR2B Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
93 | |
Research Use Only | |
ACVR2B | |
Recombinant Protein | |
Yeast | |
SGRGEAETRE CIYYNANWEL ERTNQSGLER CEGEQDKRLH CYASWRNSSG TIELVKKGCW LDDFNCYDRQ ECVATEENPQ VYFCCCEGNF CNERFTHLPE AGGPEVTYEP PPTAPTLLT | |
Greater than 90% as determined by SDS-PAGE. |