missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADA Protein
A cDNA sequence encoding the ADA was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 89170.00 SEK
Specifications
Gene ID (Entrez) | 100 |
---|---|
Name | ADA Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | ADA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15954078
|
enQuireBio™
QP10505-2UG |
2 μg |
1545.00 SEK
2µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934078
|
enQuireBio™
QP10505-10UG |
10 μg |
2405.00 SEK
10µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944078
|
enQuireBio™
QP10505-1MG |
1 mg |
89170.00 SEK
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
100 | |
Research Use Only | |
ADA | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAQTPAFDKP KVELHVHLDG SIKPETILYY GRRRGIALPA NTAEGLLNVI GMDKPLTLPD FLAKFDYYMP AIAGCREAIK RIAYEFVEMK AKEGVVYVEV RYSPHLLANS KVEPIPWNQA EGDLTPDEVV ALVGQGLQEG ERDFGVKARS ILCCMRHQPN WSPKVVELCK KYQQQTVVAI DLAGDETIPG SSLLPGHVQA YQEAVKSGIH RTVHAGEVGS AEVVKEAVDI LKTERLGHGY HTLEDQALYN RLRQENMHFE ICPWSSYLTG AWKPDTEHAV IRLKNDQANY SLNTDDPLIF KSTLDTDYQM TKRDMGFTEE EFKRLNINAA KSSFLPEDEK RELLDLLYKA YGMPPSASAG QNL | |
Greater than 85.0% as determined by SDS-PAGE. |
ADA Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity: approximately >25 units/mg. Enzymatic activity was confirmed by measuring the amount of enzyme that deaminates 1.0 umol of adenosine to inosine per minute at pH 7.5 at 25C. | |
Human | |
His | |
ADA protein 0.5 mg/ml is supplied in 20mM Tris-HCL, pH 8.0, 1mM DTT and 20% Glycerol. |