missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADA Protein
A cDNA sequence encoding the ADA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10505-10ug
Additional Details : Weight : 0.01000kg
Specifications
100 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity: approximately >25 units/mg. Enzymatic activity was confirmed by measuring the amount of enzyme that deaminates 1.0 umol of adenosine to inosine per minute at pH 7.5 at 25C. | |
Human | |
His | |
ADA protein 0.5 mg/ml is supplied in 20mM Tris-HCL, pH 8.0, 1mM DTT and 20% Glycerol. |
ADA Protein | |
Research Use Only | |
ADA | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAQTPAFDKP KVELHVHLDG SIKPETILYY GRRRGIALPA NTAEGLLNVI GMDKPLTLPD FLAKFDYYMP AIAGCREAIK RIAYEFVEMK AKEGVVYVEV RYSPHLLANS KVEPIPWNQA EGDLTPDEVV ALVGQGLQEG ERDFGVKARS ILCCMRHQPN WSPKVVELCK KYQQQTVVAI DLAGDETIPG SSLLPGHVQA YQEAVKSGIH RTVHAGEVGS AEVVKEAVDI LKTERLGHGY HTLEDQALYN RLRQENMHFE ICPWSSYLTG AWKPDTEHAV IRLKNDQANY SLNTDDPLIF KSTLDTDYQM TKRDMGFTEE EFKRLNINAA KSSFLPEDEK RELLDLLYKA YGMPPSASAG QNL | |
Greater than 85.0% as determined by SDS-PAGE. |