missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Adamts7 Protein
A DNA sequence encoding the Homo sapiens (Human) Adamts7, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8246-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_055087.2 | |
Adamts7 Protein | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
11173 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
ADAMTS7 | |
Recombinant Protein | |
Yeast | |
KWVETLVVAD AKMVEYHGQP QVESYVLTIM NMVAGLFHDP SIGNPIHITI VRLVLLEDEE EDLKITHHAD NTLKSFCKWQ KSINMKGDAH PLHHDTAILL TRKDLCAAMN RPCETLGLSH VAGMCQPHRS CSINEDTGLP LAFTVAHELG HSFGIQHDGS GNDCEPVGKR PFIMSPQLLY DAAPLTWSRC SRQYITRFLD RGWGLCLDDP PAKDIIDFPS VPPGVLYDVS HQCRLQYGAY SAFCEDMDNV CHTLWCSVGT TCHSKLDAAV DGTRCGENKW CLSGECVPVG FRPEAVDGGW SGWSAWSICS RSCGMGVQSA ERQCTQPTPK YKGRYCVGER KRFRLCNLQA CP |