missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADP-ribosylation factor-like protein 1
A DNA sequence encoding the Homo sapiens (Human) ADP-ribosylation factor-like protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8356-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001186674.1 | |
ADP-ribosylation factor-like protein 1 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
402 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
ARL2 | |
Recombinant Protein | |
E. coli | |
LLMLGLDNAG KTTILKKFNG EDIDTISPTL GFNIKTLEHR GFKLNIWDVG GQKSLRSYWR NYFESTDGLI WVVDSADRQR MQDCQRELQS LLVEERLAGA TLLIFANKQD LPGALSSNAI REVLELDSIR SHHWCIQGCS AVTGENLLPG IDWLLDDISS RIFTAD |