missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AFG3-like protein 1
A DNA sequence encoding the Homo sapiens (Human) AFG3-like protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8466-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006787.2 | |
AFG3-like protein 1 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
10939 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
AFG3L2 | |
Recombinant Protein | |
E. coli | |
ERVIGGLEKK TQVLQPEEKK TVAYHEAGHA VAGWYLEHAD PLLKVSIIPR GKGLGYAQYL PKEQYLYTKE QLLDRMCMTL GGRVSEEIFF GRITTGAQDD LRKVTQSAYA QIVQFGMNEK VGQISFDLPR QGDMVLEKPY SEATARLIDD EVRILINDAY KRTVALLTEK KADVEKVALL LLEKEVLDKN DMVELLGPRP FAEKSTYEEF |