missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AKT-interacting Protein

A DNA sequence encoding the Homo sapiens (Human) AKT-interacting protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8053-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16002391

  • 4960.00 SEK / 100µg
Estimated Shipment: 17-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001012398.1
AKT-interacting Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
64400
Greater than 90% as determined by SDS-PAGE.
Research Use Only
AKTIP
Recombinant Protein
E. coli
MNPFWSMSTS SVRKRSEGEE KTLTGDVKTS PPRTAPKKQL PSIPKNALPI TKPTSPAPAA QSTNGTHASY GPFYLEYSLL AEFTLVVKQK LPGVYVQPSY RSALMWFGVI FIRHGLYQDG VFKFTVYIPD NYPDGDCPRL VFDIPVFHPL VDPTSGELDV KRAFAKWRRN HNHIWQVLMY ARRVFYKIDT ASPLNPEAAV LYEKDIQLFK SKVVDSVKVC TARLFDQPKI EDPYAISFSP WNPSVHDEAR EKMLTQKKPE EQHNKSVHVA GLSWVKPGSV QPFSKEEKTV AT
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers