missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AN1-type zinc finger protein 1

A DNA sequence encoding the Homo sapiens (Human) AN1-type zinc finger protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8089-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062601

  • 4074.00 SEK / 100µg


Estimated Shipment
08-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_068762.1
AN1-type zinc finger protein 1
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
60685
Greater than 80% as determined by SDS-PAGE.
Research Use Only
ZFAND3
Recombinant Protein
E. coli
MGDAGSERSK APSLPPRCPC GFWGSSKTMN LCSKCFADFQ KKQPDDDSAP STSNSQSDLF SEETTSDNNN TSITTPTLSP SQQPLPTELN VTSPSKEECG PCTDTAHVSL ITPTKRSCGT DSQSENEASP VKRPRLLENT ERSEETSRSK QKSRRRCFQC QTKLELVQQE LGSCRCGYVF CMLHRLPEQH DCTFDHMGRG REEAIMKMVK LDRKVGRSCQ RIGEGCS
Videos
SDS
Documents