missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Angiopoietin-2 / ANG2 Protein
A DNA sequence encoding the Homo sapiens (Human) Angiopoietin-2 / ANG2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8512-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001112359.1 | |
Angiopoietin-2 / ANG2 Protein | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
285 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
ANGPT2 | |
Recombinant Protein | |
E. coli | |
YNNFRKSMDS IGKKQYQVQH GSCSYTFLLP EMDNCRSSSS PYVSNAVQRD APLEYDDSVQ RLQVLENIME NNTQWLMKLE NYIQDNMKKE MVEIQQNAVQ NQTAVMIEIG TNLLNQTAEQ TRKLTDVEAQ VLNQTTRLEL QLLEHSLSTN KLEKQILDQT SEINKLQDKN SFLEKKVLAM EDKHIIQLQS IKEEKDQLQV LVSKQNSIIE ELEKKIVTAT VNNSVLQKQQ HDLMETVNNL LTMMSTSNSA KDPTVAKEEQ ISFRDCAEVF KSGHTTNGIY TLTFPNSTEE IKAYCDMEAG GGGWTIIQRR EDGSVDFQRT WKEYKVGFGN PSGEYWLGNE FVSQLTNQQR YVLKIHLKDW EGNEAYSLYE HFYLSSEELN YRIHLKGLTG TAGKISSISQ PGNDFSTKDG DNDKCICKCS QMLTGGWWFD ACGPSNLNGM YYPQRQNTNK FNGIKWYYWK GSGYSLK |