missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANGPTL2 / Angiopoietin-like 2 Protein
A DNA sequence encoding the Homo sapiens (Human) ANGPTL2 / Angiopoietin-like 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8238-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036230.1 | |
ANGPTL2 / Angiopoietin-like 2 Protein | |
Research Use Only | |
ANGPTL2 | |
Recombinant Protein | |
E. coli | |
QEDGFEGTEE GSPREFIYLN RYKRAGESQD KCTYTFIVPQ QRVTGAICVN SKEPEVLLEN RVHKQELELL NNELLKQKRQ IETLQQLVEV DGGIVSEVKL LRKESRNMNS RVTQLYMQLL HEIIRKRDNA LELSQLENRI LNQTADMLQL ASKYKDLEHK YQHLATLAHN QSEIIAQLEE HCQRVPSARP VPQPPPAAPP RVYQPPTYNR IINQISTNEI QSDQNLKVLP PPLPTMPTLT SLPSSTDKPS GPWRDCLQAL EDGHDTSSIY LVKPENTNRL MQVWCDQRHD PGGWTVIQRR LDGSVNFFRN WETYKQGFGN IDGEYWLGLE NIYWLTNQGN YKLLVTMEDW SGRKVFAEYA SFRLEPESEY YKLRLGRYHG NAGDSFTWHN GKQFTTLDRD HDVYTGNCAH YQKGGWWYNA CAHSNLNGVW YRGGHYRSRY QDGVYWAEFR GGSYSLKKVV MMIRPNPNTF H | |
Greater than 90% as determined by SDS-PAGE. |
23452 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |