missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ANGPTL2 / Angiopoietin-like 2 Protein

A DNA sequence encoding the Homo sapiens (Human) ANGPTL2 / Angiopoietin-like 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8238-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16063471

  • 4074.00 SEK / 100µg


Estimated Shipment
04-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_036230.1
ANGPTL2 / Angiopoietin-like 2 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
23452
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ANGPTL2
Recombinant Protein
E. coli
QEDGFEGTEE GSPREFIYLN RYKRAGESQD KCTYTFIVPQ QRVTGAICVN SKEPEVLLEN RVHKQELELL NNELLKQKRQ IETLQQLVEV DGGIVSEVKL LRKESRNMNS RVTQLYMQLL HEIIRKRDNA LELSQLENRI LNQTADMLQL ASKYKDLEHK YQHLATLAHN QSEIIAQLEE HCQRVPSARP VPQPPPAAPP RVYQPPTYNR IINQISTNEI QSDQNLKVLP PPLPTMPTLT SLPSSTDKPS GPWRDCLQAL EDGHDTSSIY LVKPENTNRL MQVWCDQRHD PGGWTVIQRR LDGSVNFFRN WETYKQGFGN IDGEYWLGLE NIYWLTNQGN YKLLVTMEDW SGRKVFAEYA SFRLEPESEY YKLRLGRYHG NAGDSFTWHN GKQFTTLDRD HDVYTGNCAH YQKGGWWYNA CAHSNLNGVW YRGGHYRSRY QDGVYWAEFR GGSYSLKKVV MMIRPNPNTF H
Videos
SDS
Documents