missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apolipoprotein B-100 Protein
A DNA sequence encoding the Homo sapiens (Human) Apolipoprotein B-100, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8988-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000375.2 | |
Apolipoprotein B-100 Protein | |
Research Use Only | |
APOB | |
Recombinant Protein | |
Yeast | |
EEEMLENVSL VCPKDATRFK HLRKYTYNYE AESSSGVPGT ADSRSATRIN CKVELEVPQL CSFILKTSQC TLKEVYGFNP EGKALLKKTK NSEEFAAAMS | |
Greater than 90% as determined by SDS-PAGE. |
338 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |