missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apoptosis-enhancing nuclease Protein
A DNA sequence encoding the Homo sapiens (Human) Apoptosis-enhancing nuclease, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7830-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_073604.3 | |
Apoptosis-enhancing nuclease Protein | |
Research Use Only | |
AEN | |
Recombinant Protein | |
E. coli | |
MVGTGPRGRV SELARCSIVS YHGNVLYDKY IRPEMPIADY RTRWSGITRQ HMRKAVPFQV AQKEILKLLK GKVVVGHALH NDFQALKYVH PRSQTRDTTY VPNFLSEPGL HTRARVSLKD LALQLLHKKI QVGQHGHSSV EDATTAMELY RLVEVQWEQQ EARSLWTCPE DREPDSSTDM EQYMEDQYWP DDLAHGSRGG AREAQDRRN | |
Greater than 90% as determined by SDS-PAGE. |
64782 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |