missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Apoptotic protease-activating factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Apoptotic protease-activating factor 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8984-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001151.1 | |
Apoptotic protease-activating factor 1 Protein | |
Research Use Only | |
APAF1 | |
Recombinant Protein | |
Yeast | |
SGITSYVRTV LCEGGVPQRP VVFVTRKKLV NAIQQKLSKL KGEPGWVTIH GMAGCGKSVL AAEAVRDHSL LEGCFPGGVH WVSVGKQDKS GLLMKLQNLC TRLDQDESFS QRLPLNIEEA KDRLRILMLR KHPRSLLILD DVWDSWVLKA FDSQCQILLT TRDKSVTDSV MGPKYVVPVE SSLGKEKGLE ILSLFVNMKK ADLPEQAHSI IKECKGSPLV VSLIGALLRD FPNRWEYYLK QLQNKQFKRI RKSSSYDYEA LDEAMSISVE MLREDIKDYY TDLSILQKDV KVPTKVLCIL WDMETEEVED IL | |
Greater than 90% as determined by SDS-PAGE. |
317 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |