missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Aryl-hydrocarbon-interacting protein-like 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Aryl-hydrocarbon-interacting protein-like 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8180-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001028226.1 | |
Aryl-hydrocarbon-interacting protein-like 1 Protein | |
Research Use Only | |
AIPL1 | |
Recombinant Protein | |
E. coli | |
MDAALLLNVE GVKKTILHGG TGELPNFITG SRVIFHFRTM KCDEERTVID DSRQVGQPMH IIIGNMFKLE VWEILLTSMR VHEVAEFWCD TIHTGVYPIL SRSLRQMAQG KDPTEWHVHT CGLANMFAYH TLGYEDLDEL QKEPQPLVFV IELLQVDAPS DYQRETWNLS NHEKMKAVPV LHGEGNRLFK LGRYEEASSK YQEAIICLRN LQTKEKPWEV QWLKLEKMIN TLILNYCQCL LKKEEYYEVL EHTSDILRHH PGIVKAYYVR ARAHAEVWNE AEAKADLQKV LELEPSMQKA VRRELRLLEN RMAEKQEEER LRCRNMLSQG ATQPPAEPPT EPPAQSSTEP PAEPPTAPSA ELSAGPPAEP ATEPPPSPGH SLQH | |
Greater than 80% as determined by SDS-PAGE. |
23746 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |