missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ASH2L Protein

A DNA sequence encoding the Homo sapiens (Human) ASH2L, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8184-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013131

  • 5980.00 SEK / 100µg
Estimated Shipment: 10-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001098684.1
ASH2L Protein
Research Use Only
ASH2L
Recombinant Protein
E. coli
MDTQAGSVDE ENGRQLGEVE LQCGICTKWF TADTFGIDTS SCLPFMTNYS FHCNVCHHSG NTYFLRKQAN LKEMCLSALA NLTWQSRTQD EHPKTMFSKD KDIIPFIDKY WECMTTRQRP GKMTWPNNIV KTMSKERDVF LVKEHPDPGS KDPEEDYPKF GLLDQDLSNI GPAYDNQKQS SAVSTSGNLN GGIAAGSSGK GRGAKRKQQD GGTTGTTKKA RSDPLFSAQR LPPHGYPLEH PFNKDGYRYI LAEPDPHAPD PEKLELDCWA GKPIPGDLYR ACLYERVLLA LHDRAPQLKI SDDRLTVVGE KGYSMVRASH GVRKGAWYFE ITVDEMPPDT AARLGWSQPL GNLQAPLGYD KFSYSWRSKK GTKFHQSIGK HYSSGYGQGD VLGFYINLPE DTETAKSLPD TYKDKALIKF KSYLYFEEKD FVDKAEKSLK QTPHSEIIFY KNGVNQGVAY KDIFEGVYFP AISLYKSCTV SINFGPCFKY PPKDLTYRPM SDMGWGAVVE HTLADVLYHV ETEVDGRRSP PWEP
Greater than 90% as determined by SDS-PAGE.
9070
100 μg
Not determined.
Full Length of Isoform 3
Human
His-SUMO
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers