missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BANK1 Protein
A DNA sequence encoding the Homo sapiens (Human) BANK1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9855-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001077376.2 | |
BANK1 Protein | |
Research Use Only | |
BANK1 | |
Recombinant Protein | |
Yeast | |
MLPAAPGKGL GSPDPAPCGP APPGNTKDII MIYEEDAEEW ALYLTEVFLH VVKREAILLY RLENFSFRHL ELLNLTSYKC KLLILSNSLL RDLTPKKCQF LEKILHSPKS VVTLLCGVKS SDQLYELLNI SQSRWEISTE QEPEDYISVI QSIIFKDSED YFEVNIPTDL RAKHSGEISE RKEIEELSEA SRNTIPLAVV LPTEIPCENP GEIFIILRDE VIGDTVEVEF TSSNKRIRTR PALWNKKVWC MKALEFPAGS VHVNVYCDGI VKATTKIKYY PTAKAKECLF RMADSGESLC QNSIEELDGV LTSIFKHEIP YYEFQSLQTE ICSQNKYTHF KELPTLLHCA AKFGLKNLAI HLLQCSGATW ASKMKNMEGS DPAHIAERHG HKELKKIFED FSIQEIDINN EQENDYEEDI ASFSTYIPST QNPAFHHESR KTYGQSADGA EANEMEGEGK QNGSGMETKH SPLEVGSESS EDQYDDLYVF IPGADPENNS QEPLMSSRPP LPPPRPVANA FQLERPHFTL PGTMVEGQME RSQNWGHPGV RQETGDEPKG EKEKKEEEKE QEEEEDPYTF AEIDDSEYDM ILANLSIKKK TGSRSFIINR PPAPTPRPTS IPPKEETTPY IAQVFQQKTA RRQSDDDKFC GLPKKQDRAR IESPAFSTLR GCLTDGQEEL ILLQEKVKNG KMSMDEALEK FKHWQMGKSG LEMIQQEKLR QLRDCIIGKR PEEENVYNKL TIVHHPGGKE TAHNENKFYN VHFSNKLPAR PQVEKEFGFC CKKDH | |
Greater than 90% as determined by SDS-PAGE. |
55024 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |