missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BANK1 Protein
A DNA sequence encoding the Homo sapiens (Human) BANK1, was expressed in the hosts and tags indicated.
2075.00 SEK - 22323.03 SEK
Specifications
Name | BANK1 Protein |
---|---|
Product Type | Recombinant Protein |
Protein Tag | His |
Host Species | Yeast |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16058851
|
enQuireBio™
QP9855-YE-100UG |
100 μg |
5095.55 SEK
100µg |
Estimated Shipment 19-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! |
16078851
|
enQuireBio™
QP9855-YE-1MG |
1 mg |
22323.03 SEK
1mg |
Estimated Shipment 19-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! |
16068851
|
enQuireBio™
QP9855-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
16088851
|
enQuireBio™
QP9855-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
16098851
|
enQuireBio™
QP9855-YE-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
16008861
|
enQuireBio™
QP9855-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
Specifications
BANK1 Protein | |
His | |
Human | |
Greater than 90% as determined by SDS-PAGE. | |
NP_001077376.2 |
Recombinant Protein | |
Yeast | |
MLPAAPGKGL GSPDPAPCGP APPGNTKDII MIYEEDAEEW ALYLTEVFLH VVKREAILLY RLENFSFRHL ELLNLTSYKC KLLILSNSLL RDLTPKKCQF LEKILHSPKS VVTLLCGVKS SDQLYELLNI SQSRWEISTE QEPEDYISVI QSIIFKDSED YFEVNIPTDL RAKHSGEISE RKEIEELSEA SRNTIPLAVV LPTEIPCENP GEIFIILRDE VIGDTVEVEF TSSNKRIRTR PALWNKKVWC MKALEFPAGS VHVNVYCDGI VKATTKIKYY PTAKAKECLF RMADSGESLC QNSIEELDGV LTSIFKHEIP YYEFQSLQTE ICSQNKYTHF KELPTLLHCA AKFGLKNLAI HLLQCSGATW ASKMKNMEGS DPAHIAERHG HKELKKIFED FSIQEIDINN EQENDYEEDI ASFSTYIPST QNPAFHHESR KTYGQSADGA EANEMEGEGK QNGSGMETKH SPLEVGSESS EDQYDDLYVF IPGADPENNS QEPLMSSRPP LPPPRPVANA FQLERPHFTL PGTMVEGQME RSQNWGHPGV RQETGDEPKG EKEKKEEEKE QEEEEDPYTF AEIDDSEYDM ILANLSIKKK TGSRSFIINR PPAPTPRPTS IPPKEETTPY IAQVFQQKTA RRQSDDDKFC GLPKKQDRAR IESPAFSTLR GCLTDGQEEL ILLQEKVKNG KMSMDEALEK FKHWQMGKSG LEMIQQEKLR QLRDCIIGKR PEEENVYNKL TIVHHPGGKE TAHNENKFYN VHFSNKLPAR PQVEKEFGFC CKKDH | |
55024 | |
BANK1 |