missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Bifunctional epoxide hydrolase 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Bifunctional epoxide hydrolase 2, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001243411.1 |
---|---|
Gene ID (Entrez) | 2053 |
Name | Bifunctional epoxide hydrolase 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15989149
|
enQuireBio™
QP5986-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15909159
|
enQuireBio™
QP5986-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999149
|
enQuireBio™
QP5986-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15919159
|
enQuireBio™
QP5986-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15929159
|
enQuireBio™
QP5986-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15939159
|
enQuireBio™
QP5986-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001243411.1 | |
Bifunctional epoxide hydrolase 2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
2053 | |
Research Use Only | |
EPHX2 | |
Recombinant Protein | |
E. coli | |
MTLRAAVFDL DGVLALPAVF GVLGRTEEAL ALPRGLLNDA FQKGGPEGAT TRLMKGEITL SQWIPLMEEN CRKCSETAKV CLPKNFSIKE IFDKAISARK INRPMLQAAL MLRKKGFTTA ILTNTWLDDR AERDGLAQLM CELKMHFDFL IESCQVGMVK PEPQIYKFLL DTLKASPSEV VFLDDIGANL KPARDLGMVT ILVQDTDTAL KELEKVTGIQ LLNTPAPLPT SCNPSDMSHG YVTVKPRVRL HFVELGSGPA VCLCHGFPES WYSWRYQIPA LAQAGYRVLA MDMKGYGESS APPEIEEYCM EVLCKEMVTF LDKLGLSQAV FIGHDWGGML VWYMALFYPE RVRAVASLNT PFIPANPNMS PLESIKANPV FDYQLYFQEP GVAEAELEQN LSRTFKSLFR ASDESVLSMH KVCEAGGLFV NSPEEPSLSR MVTEEEIQFY VQQFKKSGFR GPLNWYRNME RNWKWACKSL GRKILIPALM VTAEKDFVLV PQMSQHMEDW IPHLKRGHIE DCGHWTQMDK PTEVNQILIK WLDSDARNPP VVSKM | |
Greater than 90% as determined by SDS-PAGE. |