missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Butyrophilin subfamily 2 member A1 Protein
A DNA sequence encoding the Homo sapiens (Human) Butyrophilin subfamily 2 member A1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9818-ye-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001184162.1 | |
Butyrophilin subfamily 2 member A1 Protein | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
11120 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
BTN2A1 | |
Recombinant Protein | |
Yeast | |
QFIVVGPTDP ILATVGENTT LRCHLSPEKN AEDMEVRWFR SQFSPAVFVY KGGRERTEEQ MEEYRGRTTF VSKDISRGSV ALVIHNITAQ ENGTYRCYFQ EGRSYDEAIL HLVVAGLGSK PLISMRGHED GGIRLECISR GWYPKPLTVW RDPYGGVAPA LKEVSMPDAD GLFMVTTAVI IRDKSVRNMS CSINNTLLGQ KKESVIFIPE SFMPSVSPCA |