missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Butyrophilin subfamily 2 member A2 Protein

A DNA sequence encoding the Homo sapiens (Human) Butyrophilin subfamily 2 member A2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7953-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16061811

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001184166.1
Butyrophilin subfamily 2 member A2 Protein
50 μg
Not determined.
Partial (see sequence information for more details). of Isoform 2
Human
His-SUMO
Tris-based buffer, 50% glycerol
10385
Greater than 90% as determined by SDS-PAGE.
Research Use Only
BTN2A2
Recombinant Protein
E. coli
QFTVVGPANP ILAMVGENTT LRCHLSPEKN AEDMEVRWFR SQFSPAVFVY KGGRERTEEQ MEEYRGRITF VSKDINRGSV ALVIHNVTAQ ENGIYRCYFQ EGRSYDEAIL RLVVAGLGSK PLIEIKAQED GSIWLECISG GWYPEPLTVW RDPYGEVVPA LKEVSIADAD GLFMVTTAVI IRDKYVRNVS CSVNNTLLGQ EKETVIFIPE SFMPSASPWM VALAVILTAS
Videos
SDS
Documents